Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ISG15/UCRP, Clone: 851701, R&D Systems™

Catalog No. A830
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A830 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A830 Supplier R&D Systems Supplier No. A830

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

ISG15/UCRP Monoclonal specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ISG15/UCRP
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 851701
Conjugate Unconjugated
Dilution Western Blot 0.1-0.5 ug/mL, Immunoprecipitation 10-20 ug/mL
Formulation Supplied as a solution in PBS containing Glycerol and Sodium Azide. with Sodium Azide
Gene Accession No. P05161
Gene Alias G1P2, G1P2interferon-induced 17-kDa/15-kDa protein, IFI15, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, IP17, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRP, UCRPIP17
Gene Symbols ISG15
Host Species Rat
Immunogen Full-length recombinant human ISG15/UCRP Accession # P05161
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9636
Test Specificity This antibody will recover ISGylated proteins in immunoprecipitation applications. This antibody detects ISGylated proteins in Western blots.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.