Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISG15/UCRP Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 09412MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-412-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-412-MI Supplier MilliporeSigma Supplier No. 09412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect ISG15/UCRP using this Anti-ISG15/UCRP Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISG15/UCRP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 100μL of purified by salt precipitation and peptide immunochromatography in PBS containing 0.01M sodium azide. Dilute to working strength with phosphate buffered saline pH 7.2-7.4 and 1% normal goat serum (if a goat anti-rabbit IgG linker antibody is
Gene Symbols ISG15; UCRP; IFI15; G1P2
Host Species Rabbit
Immunogen Peptide corresponding to residues 85-101 of human ISG15.
Purification Method Salt precipitation
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_005101.2
Target Species Human
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.