Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Islet-1 Mouse, Unlabeled, Clone: Q11-465, BD

Catalog No. BDB562546 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562546 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562546 Supplier BD Biosciences Supplier No. 562546
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Islet-1 is a LIM-homeodomain transcription factor important for motor neuron differentiation and the formation of islet cells in the pancreas. Various heart cell types, such as cardiac muscle, the conduction system and endothelial cells in multiple heart tissue compartments during cardiogenesis, have been found to originate from Islet-1-positive cardiac precursor cells. Moreover, Islet-1-positive cells from differentiated human embryonic stem cell lines were found to be capable of self-renewal and expansion and could differentiate into the three major cell types of the heart. Western blot analyses of lysates from transfected cells demonstrate that the Q11-465 monoclonal antibody reacts with human Islet-1 and Islet-2, similar to the 4D5 clone (Tsuchida et al, 1994). Cross-reactivity with mouse Islet is also observed.

  • Alternate Name: ISL-1, ISL1, Islet-2, ISL-2, ISL2
  • Isotype: IgG1, κ
  • Host Species: Mouse
  • Cross Reactivity: Mouse, Human
  • Immunogen: Human Islet-1 Recombinant Protein
  • Target MW: 45-50kDa
  • Unlabeled

Western Blotting, Bioimaging, Immunofluorescence, Intracellular Staining (Flow Cytometry)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Islet-1
Applications Western Blot
Classification Monoclonal
Clone Q11-465
Concentration 0.5mg/mL
Conjugate Unconjugated
Description ISL-1, ISL1, Islet-2, ISL-2, ISL2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Islet-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.