Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITFG1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA531872
Description
PA5-31872 targets ITFG1 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-31872 immunogen is recombit fragment corresponding to a region within amino acids 17 and 301 of Human ITFG1.
Modulator of T-cell function. Has a protective effect in graft versus host disease model.
Specifications
ITFG1 | |
Polyclonal | |
Unconjugated | |
Q8TB96 | |
ITFG1 | |
Recombit fragment corresponding to a region within amino acids 17 and 301 of Human ITFG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.53 mg/mL | |
ITFG1 | |
TIP, 2310047C21Rik, T cell immunomodulatory protein, T-cell immunomodulatory protein, integrin-alpha FG-GAP repeat-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81533 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction