Learn More
Description
Specifications
Specifications
| Antigen | Itk/Emt/Tsk |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polycloncal in buffer containing PBS with 0.05% sodium azide and 50% glycerol. |
| Gene Symbols | EC 2.7.10.2 ; EMT ; LYK; MGC126257; MGC126258; PSCTK2 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein. Itk/Emt/Tsk-GST fusion protein (Itk/Emt/Tsk amino acid residues 1-270) spanning the PH SH3 domains and a small portion of the SH2 domain; bound to nitrocellulose paper. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.