Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

inositol 1,4,5-trisphosphate 3-kinase B, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017795 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-795 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-795 Supplier Abnova Corporation Supplier No. H00003707D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human ITPKB protein.

The protein encoded by this protein regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of this encoded protein is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. [provided by RefSeq

Sequence: MAVYCYALNSLVIMNSANEMKSGGGPGPSGSETPPPPRRAVLSPGSVFSPGRGASFLFPPAESLSPEEPRSPGGWRSGRRRLNSSSGSGSGSSGSSVSSPSWAGRLRGDRQQVVAAGTLSPPGPEEAKRKLRILQRELQNVQVNQKVGMFEAHIQAQSSAIQAPRSPRLGRAHSPSPCPFRSSSQPPGRVLVQGARSEERRTKSWGEQCPETSGTDSGRKGGPSLCSSQVKKGMPPLPGRAAPTGSEAQGPSAFVRMEKGIPASPRCGSPTAMEIDKRGSPTPGTRSCLAPSLGLFGASLTMATEVAARVTSTGPHRPQDLALTEPSGRARELEDLQPPEALVERQGQFLGSETSPAPERGGPRDGEPPGKMGKGYLPCGMPGSGEPEVGKRPEETTVSVQSAESSDALSWSRLPRALASVGPEEARSGAPVGGGRWQLSDRVEGGSPTLGLLGGSPSAQPGTGNVEAGIPSGRMLEPLPCWDAAKDLKEPQCPPGDRVGVQPGNSRVWQGTMEKAGLAWTRGTGVQSEGTWESQRQDSDALPSPELLPQDQDKPFLRKACSPSNIPAVIITDMGTQEDGALEETQGSPRGNLPLRKLSSSSASSTGFSSSYEDSEEDISSDPERTLDPNSAFLHTLDQQKPRV

Specifications

Antigen inositol 1,4,5-trisphosphate 3-kinase B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ITPKB protein.
Formulation PBS with no preservative; pH 7.4
Gene ITPKB
Gene Accession No. BC015009.1
Gene Alias IP3K/IP3K-B/IP3KB/PIG37
Gene Symbols ITPKB
Host Species Rabbit
Immunogen ITPKB (AAH15009.1, 1 a.a. ∼ 644 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3707
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.