Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
J Chain Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA532457
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at roo
J chain is a small glycopeptide with MW of 15kDa. It is structurally unrelated to heavy or light chains, but is synthesized by all plasma cells that secrete polymeric immunoglobulins. J chain is linked to IgA and IgM by disulfide bonds, however, it has been detected in IgG- and IgD-containing cells. J chains are present in a large proportion of the immunoglobulin-positive cells in the germinal centers of the tonsils and lymph nodes. B cells discrete J chain at an early stage of differentiation with the expression persisting in those cells destined to produce IgA or IgM.
Specifications
J Chain | |
Polyclonal | |
JCHAIN | |
IGCJ, JCH, IgJ chain, immunoglobulin J chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3512 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P01591 | |
JCHAIN | |
Synthetic peptide derived from near C-terminus of human J-chain protein. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction