Learn More
Description
The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. The protein encoded by this gene is one of several ligands that activate Notch and related receptors. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | jagged 2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1C5 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant JAG2. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | JAG2 |
| Gene Accession No. | NM_002226 |
| Gene Alias | HJ2/SER2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.