Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Jagged 1/Jagged 2, PE, Clone: 241002, R&D Systems™

Catalog No. FAB1726P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1726P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1726P Supplier R&D Systems Supplier No. FAB1726P

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Jagged 1/Jagged 2 Monoclonal specifically detects Jagged 1/Jagged 2 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Jagged 1/Jagged 2
Applications Flow Cytometry
Classification Monoclonal
Clone 241002
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9Y219
Gene Symbols JAG1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Jagged 2 Met27-Asp307 Accession # Q9Y219
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 182
Test Specificity Stains human Jagged 1 and Jagged 2 transfectants but not the parental cell line by flow cytometry. Detects endogenous human and mouse Jagged 1 and Jagged 2 by flow cytometry.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.