Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071485MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-148-5MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-148-5MI Supplier MilliporeSigma Supplier No. 071485

Rabbit Polyclonal Antibody

Detect JAK1 using this Anti-JAK1 Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in 0.02M Phosphate with 0.15M NaCl, pH 7.6 and 0.1% sodium azide as a preservative.
Gene Symbols JAK1; JAK1B; JAK-1; JAK1A; EC 2.7.10.2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 1123-1141 of human JAK1 (CQPSNRTSFQNLIEGFEALLK)
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002227.2
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.