Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK1 Rabbit anti-Human, Hamster, Mouse, Rat, Rabbit, Polyclonal, MilliporeSigma™

Catalog No. 06272MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-272-MI 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-272-MI Supplier MilliporeSigma Supplier No. 06272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-JAK1 Antibody detects level of JAK1 and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAK1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation whole rabbit antiserum containing 0.05% sodium azide
Gene Symbols JAK1; JAK1B; JAK1A; JAK-1
Host Species Rabbit
Immunogen HPLC-purified (≥98%) peptide corresponding to 21 amino acids of human JAK1 (KTLIEKERFYESRCRPVTPS-C)
Purification Method Antiserum
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002227
Target Species Hamster, Human, Mouse, Rabbit, Rat
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.