Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK1 Mouse, Unlabeled, Clone: 73, BD

Catalog No. BDB610231 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610231 50 μg
BDB610232 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610231 Supplier BD Biosciences Supplier No. 610231
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The JAK family of receptor-associated protein kinases is directly involved in interferon (IFN) response pathways. The JAK family contains at least three members: JAK1, JAK2, and Tyk2. Each protein is approximately 130kDa and contains a C-terminal tyrosine kinase domain, an adjacent kinase or kinase-related domain, and five other domains that are highly conserved among family members. In several human and murine cell lines, JAK1 is rapidly tyrosine phosphorylated in response to IFN-α and IFN-γ. JAK1 is required for the phosphorylation of the transcription factor Stat1 (p91), in response to IFNs-α or IFN-γ. JAK1 is also necessary for the efficient phophorylation of Stat2 (p113) in response to IFN-α and for the phosphorylation of Tyk2 or JAK2 in response to IFNs-α or IFN-γ, respectively.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen JAK1
Applications Western Blot
Classification Monoclonal
Clone 73
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.