Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK1 Mouse anti-Human, Mouse, Rat, Canine, Xenopus, Chicken, Clone: 73, MilliporeSigma™

Catalog No. 051154MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-115-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-115-4MI Supplier MilliporeSigma Supplier No. 051154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-JAK1 Antibody, clone 73 detects level of JAK1 and has been published and validated for use in western blot, immunoprecipitation and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAK1
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 73
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2b aqueous buffered solution containing 50% glycerol, BSA, and < 0.09% sodium azide
Gene Accession No. NM_002227.2
Gene Symbols JAK1, JAK1B, JAK-1, JAK1A, EC 2.7.10.2
Host Species Mouse
Immunogen Generated from human JAK1, amino acids 551-776.
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat, Xenopus species
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.