Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK2 Rabbit anti-Mouse, Human, Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 061310MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-131-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-131-0MI Supplier MilliporeSigma Supplier No. 061310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-JAK2 Antibody is validated for use in western blot, immunoprecipitation for the detection of JAK2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAK2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols JAK2; JAK-2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the protein kinase 1 domain of mouse JAK2.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001041642
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.