Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 16121
Description
Detect JAK2 using this Anti-JAK2 Antibody, agarose conjugate validated for use in immunoprecipitation.
Specifications
JAK2 | |
Polyclonal | |
200μg of Anti-JAK2 covalently bound to 200μL settled Protein A agarose; provided as a 50% gel slurry for a total volume of 400μL, suspended in phosphate-buffered saline (PBS) containing 0.05% sodium azide. Liquid suspension. | |
Rabbit | |
200 μg | |
NM_004972.2 | |
4°C for 12 months |
Immunoprecipitation | |
Agarose | |
JAK2; OTTHUMP00000043260; JAK-2 | |
murine JAK2 residues 758-776 | |
Primary | |
Human, Mouse, Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction