Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-JAK2, Polyclonal

Catalog No. 06255IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
06-255-IMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-255-IMI Supplier MilliporeSigma™ Supplier No. 06255I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects JAK2 Clone: in Human, Mouse, Rat samples, and it is validated for Immunoprecipitation, Western Blotting

Tyrosine-protein kinase JAK2 (EC 2.7.10.2; UniProt Q62120; also known as JAK-2, Janus kinase 2) is encoded by the Jak2 (also known as Fd17) gene (Gene ID 16452) in murine species. JAK2 belongs to the family of non-receptor Janus tyrosine kinases that regulate a spectrum of cellular functions downstream of activated cytokine receptors in the lympho-hematopoietic system. Immunological stimuli, such as interferons and cytokines cause the recruitment of Stat transcription factors to cytokine receptor. Subsequent phosphorylation by receptor-associated JAK2 causes Stat dimerization, nuclear translocation, and Stat-mediated target genes transcription. The canonical JAK/Stat pathway is integral to maintaining a normal immune system by stimulating proliferation, differentiation, survival, and host resistance to pathogens. Altering JAK/Stat signaling to reduce immune cells cytokine responses represents an attractive target for anti-inflammatory therapies. Within the JAK2 kinase domain, there is a region that has considerable sequence homology to the regulatory region of the insulin receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAK2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q62120
Gene Symbols Jak2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to residues 758-776 of mouse JAK2.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001041642
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.