Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK3 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. 06342MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-342-MI 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-342-MI Supplier MilliporeSigma Supplier No. 06342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-JAK3 Antibody is an antibody against JAK3 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAK3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation antiserum containing 0.05% sodium azide
Gene Symbols JAK3; LJAK; L-JAK; JAK-3; JAKL; JAK3_HUMAN
Host Species Rabbit
Immunogen peptide corresponding to residues 934-952 (RDELCVAGAQLYACQDPAI) of murine JAK3. The immunizing peptide sequence shares 16/19 amino acids with rat JAK3, but contains an amino acid insertion, which may block cross-reactivity. The immunizing peptide shares 1
Purification Method Antiserum
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000215
Target Species Mouse
Content And Storage −20°C for 24 months. Aliquot to avoid repeated freezing and thawing
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.