Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK3 Mouse anti-Human, Mouse, Clone: B32-32, MilliporeSigma™

Catalog No. 05406MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-406-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-406-MI Supplier MilliporeSigma Supplier No. 05406
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-JAK3 Antibody, clone B32-32 detects level of JAK3 and has been published and validated for use in immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAK3
Applications Immunoprecipitation
Classification Monoclonal
Clone B32-32
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl containing 0.05% sodium azide
Gene Accession No. NM_000215
Gene Symbols JAK3, LJAK, L-JAK, JAK-3, JAKL, JAK3_HUMAN
Host Species Mouse
Immunogen GST-fusion protein containing amino acids 1076-1100 of murine thymus JAK3. Clone B32-32.
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.