Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAM-A Rat anti-Mouse, Clone: BV11, MilliporeSigma™

Catalog No. MABT128MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT128MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT128MI Supplier MilliporeSigma Supplier No. MABT128
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect JAM-A using this Anti-JAM-A Antibody, clone BV11 validated for use in flow cytometry, IC and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAM-A
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone BV11
Conjugate Unconjugated
Formulation Purified rat monocolonal IgG2bκ in buffer containing PBS and no preservatives.
Gene Accession No. NP_766235
Gene Symbols F11r, Jam1, Jcam, Jcam1
Host Species Rat
Immunogen H5V murine heart capillary endothelial cells.
Purification Method Protein G Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.