Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-JAM1 (NT), Clone: EP1042Y, Rabbit Monoclonal,

Catalog No. 04593MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-593-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-593-MI Supplier MilliporeSigma™ Supplier No. 04593

Specifications

Antigen JAM1 (NT)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone EP1042Y
Conjugate Unconjugated
Formulation Buffer: 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA.
Gene Accession No. NM_144502.1, NM_144504.1, NM_016946.3, NM_144503.1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues near the N-terminus of human JAM1.
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) F11R, JAM-A, PAM-1, JAMA, CD321, JAM, JAM1, JAM-1, JCAM, KAT
Target Species Human, Mouse, Rat
Content And Storage −20°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.