Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAML Armenian Hamster anti-Mouse, PE, Clone: 4E10 (also known as HL4E10), BD

Catalog No. BDB564218 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564218 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564218 Supplier BD Biosciences Supplier No. 564218

Armenian Hamster Monoclonal Antibody

The 4E10 monoclonal antibody specifically binds to Junctional Adhesion Molecule-Like (JAML) which is also known as JAML1, AMICA, Amica1, Crea7, mCrea7, and Gm638. JAML is a type I transmembrane glycoprotein whose extracellular region contains two immunoglobulin domains. The 4E10 antibody binds to the membrane-proximal D2 Ig domain of JAML which is expressed on g ranulocytes, monocytes, memory T cells and γδ T cells. JAML serves as an endogenous ligand for epithelial and endothelial Coxsackie and Adenovirus Receptor (CAR). The JAML-CAR interaction supports leucocyte migration and the costimulation of TCR-triggered epithelial γδ T cells. The 4E10 antibody can induce costimulatory signals through JAML leading to increased activation, cytokine production and proliferation by epithelial γδ T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen JAML
Applications Flow Cytometry
Classification Monoclonal
Clone 4E10 (also known as HL4E10)
Concentration 0.2mg/mL
Conjugate PE
Description JAML1; AMICA; Amica1; Crea7; mCrea7; Gm638
Host Species Armenian Hamster
Immunogen Mouse Skin γδ T Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.