Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAMP Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | JAMP |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP236446SS
![]() |
Novus Biologicals
NBP236446SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP236446
![]() |
Novus Biologicals
NBP236446 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
JAMP Polyclonal specifically detects JAMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| JAMP | |
| Western Blot 2 ug/ml, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Unfolded Protein Response (UPR) | |
| PBS with 0.05% Sodium Azide | |
| C14orf100, chromosome 14 open reading frame 100, HSPC213, HSPC327, JAMPCDA06, JNK1/MAPK8-associated membrane protein, JNK1-associated membrane protein, JNK-associated membrane protein, Jun N-terminal kinase 1-associated membrane protein, Medulloblastoma antigen MU-MB-50.4 | |
| JKAMP | |
| IgG | |
| Affinity Purified |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9P055 | |
| 51528 | |
| A synthetic peptide made to an internal portion of the human JAMP protein (between amino acids 20-150) [UniProt Q9P055] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title