Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JARID1C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529053
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (99%), Dog (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Serine/threonine kinase which acts as a negative regulator of Ras-related Rap2-mediated signal transduction to control neuronal structure and AMPA receptor trafficking. Required for normal synaptic density, dendrite complexity, as well as surface AMPA receptor expression in hippocampal neurons. Can activate the JNK and MAPK14/p38 pathways and mediates stimulation of the stress-activated protein kinase MAPK14/p38 MAPK downstream of the Raf/ERK pathway. Phosphorylates: TANC1 upon stimulation by RAP2A, MBP and SMAD1. Has an essential function in negative selection of thymocytes, perhaps by coupling NCK1 to activation of JNK1.; Isoform 4 can activate the JNK pathway. Involved in the regulation of actin cytoskeleton reorganization, cell-matrix adhesion, cell-cell adhesion and cell migration.
Specifications
JARID1C | |
Polyclonal | |
Unconjugated | |
KDM5C | |
D930009K15Rik; DXS1272E; Histone demethylase JARID1C; JARID1C; JmjC domain-containing protein SMCX; jumonji, AT rich interactive domain 1C (Rbp2 like); Jumonji, AT rich interactive domain 1C (RBP2-like); Jumonji/ARID domain-containing protein 1C; KDM5C; Kiaa0234; lysine (K)-specific demethylase 5C; lysine demethylase 5C; lysine-specific demethylase 5C; mKIAA0234; MRX13; MRXJ; MRXSCJ; MRXSJ; Protein SmcX; Protein Xe169; RGD1560601; sele; selected mouse cDNA on the X; SMCX; Smcx homolog, X chromosome; Smcy homolog, X-linked; XE169 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8242 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P41229 | |
KDM5C | |
Recombinant fragment corresponding to a region within amino acids 10 and 291 of Human JARID1C. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction