Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAZF zinc finger 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016632 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-632 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-632 Supplier Abnova Corporation Supplier No. H00221895B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human JAZF1 protein.

This gene encodes a nuclear protein with three C2H2-type zinc fingers, and functions as a transcriptional repressor. Chromosomal aberrations involving this gene are associated with endometrial stromal tumors. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized [provided by RefSeq

Sequence: MTGIAAASFFSNTCRFGGCGLHFPTLADLIEHIEDNHIDTDPRVLEKQELQQPTYVALSYINRFMTDAARREQESLKKKIQPKLSLTLSSSVSRGNVSTPPRHSSGSLTPPVTPPITPSSSFRSSTPTGSEYGEEEVDYEESDSDESWTTESAISSEAILSSMCMNGGEEKPFACPVPGCKKRYKNVNGIKYHAKNGHRTQIRVRKPFKCRCGKSYKTAQGLRHHTINFHPPVSAEIIRKMQQ

Specifications

Antigen JAZF zinc finger 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human JAZF1 protein.
Formulation PBS with no preservative; pH 7.4
Gene JAZF1
Gene Accession No. BC042441.1
Gene Alias DKFZp761K2222/TIP27/ZNF802
Gene Symbols JAZF1
Host Species Mouse
Immunogen JAZF1 (AAH42441.1, 1 a.a. ∼ 243 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221895
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.