Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JHDM1B Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 09864MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-864-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-864-MI Supplier MilliporeSigma Supplier No. 09864
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-JHDM1B Antibody is a rabbit polyclonal antibody for detection of JHDM1B also known as lysine (K)-specific demethylase 2B, F-box protein FBL10, [Histone-H3]-lysine-36 demethylase 1B and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JHDM1B
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols KDM2B; CXXC2; FBXL10; JHDM1B; PCCX2; Fbl10; FBL10
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human JHDM1B.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_115979
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.