Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JIP-1 Mouse, Unlabeled, Clone: 50, BD

Catalog No. BDB611890 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611890 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611890 Supplier BD Biosciences Supplier No. 611890

Mouse Monoclonal Antibody

The JNK group of MAPKs is activated by a variety of inflammatory cytokines and environmental stressors. Activated JNK phosphorylates many cellular proteins, such as components of the AP-1 transcription factor complex (c-Jun and ATF-2). JNK-interacting proteins (JIPs) bind JNK, MKK7, MLKs, p190RhoGEF, and the Ste20-related protein kinase HPK1. In mouse, alternative splicing of JIP produces multiple splice variants, JIP-1, JIP-1b, JIP-2a, JIP-2b, and JIP-3. The structure of full length JIP consists of two N-terminal acidic regions, a JNK binding domain (JBD), two proline rich regions (PR), and both an SH3 and a phosphotyrosine-binding domain in the C-terminal region. JIP-1 localizes to the tips of neurites in differentiated PC12 cells, and may interact with JNK, MKK7, and MLK to facilitate formation of the JNK activating complex. IB1 is the rat homologue of JIP-1b, a JIP-1 variant that includes a 47 amino acid insert in the C-terminal region. IB1 is found in the nucleus and cytoplasm, and may function as a transactivator of the GLUT2 gene. Thus, JIP-1 and its related isoforms may have multiple functions that involve specific protein-protein interactions.

Host Species: Mouse
Clone: 50
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse JIP-1 aa. 180-384

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen JIP-1
Applications Western Blot
Classification Monoclonal
Clone 50
Concentration 250μg/mL
Conjugate Unconjugated
Description JNK Interacting Protein
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse JIP-1 aa. 180-384
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.