Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD1B Rabbit anti-Human, Mouse, Rat, Rabbit, Rhesus Macaque, Horse, Polyclonal, MilliporeSigma™

Catalog No. 09816MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-816-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-816-MI Supplier MilliporeSigma Supplier No. 09816

Rabbit Polyclonal Antibody

Anti-JMJD1B Antibody is a rabbit polyclonal antibody for detection of JMJD1B also known as Jumonji domain-containing protein 1B, Nuclear protein 5qNCA, chromosome 5 open reading frame 7 and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD1B
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols 5qNCA; JHDM2B; KDM3B; JMJD1B
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human JMJD1B.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_057688
Target Species Equine, Human, Mouse, Rabbit, Rat, Rhesus Monkey
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.