Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNK/SAPK (pT183/pY185) Mouse, Unlabeled, Clone: 41, BD

Catalog No. BDB612541 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612541 150 μg
BDB612540 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612541 Supplier BD Biosciences Supplier No. 612541
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Ras signaling pathway links the signals from growth factor receptors with the activation of the MAPK kinase cascade of phosphorylation leading to cell growth and differentiation. External stimuli, like endotoxins, UV irradiation, heat, and hyperosmolarity, induce an array of cellular responses that culminate with gene expression, ultimately dictating an adaptation to the new environment. Small GTPases of the Rho family, including cdc42, Rac1, and Rho, transmit the stress signals that initiate the signal cascade. JNK is a c-Jun kinase that was also identified as SAPK1 and MAPKp49. JNK/SAPK, along with p38 and RK5/BMK1, comprise three classes of stress-activated MAPK groups. Complete activation of JNK/SAPK requires the phosphorylation of both Thr183 and Tyr185, which are located in a Thr-X-Tyr motif. The activation of these residues is believed to be carried out by MKK4 and MKK7. Active JNK/SAPK phosphorylates other kinases and multiple transcription factors that induce expression of genes, such as proinflammatory cytokines.

Flow Cytometry, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK/SAPK (pT183/pY185)
Applications Western Blot
Classification Monoclonal
Clone 41
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human JNK/SAPK (pT183/pY185) Peptide
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.