Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNK/SAPK1 Mouse, Unlabeled, Clone: 37, BD

Catalog No. BDB610627 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610627 50 μg
BDB610628 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610627 Supplier BD Biosciences Supplier No. 610627
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Ras signaling pathway links the signals from growth factor receptors with the activation of the MAPK kinase cascade of phosphorylation leading to cell growth and differentiation. External stimuli, such as endotoxins, UV irradiation, heat, and hyperosmolarity, induce an array of cellular responses that culminate with gene expression, ultimately dictating an adaptation to the new environment. The ultimate cellular response to environmental stress is cell death or apoptosis. This pathway is regulated by Rac1 and Cdc42 which link Jun Kinases (JNKs), such as p38. JNK is a c-Jun kinase that was also identified as SAPK1 and MAPKp49. JNK contains homology with the MAP/ERK family of kinases, and contains an N-terminal ATP-binding site and two autophosphorylation “TXY” sites. JNKK1/MEKK4/SEK1 is a dual-specificity kinase activated by conditions of cellular stress, such as heat shock and hyperosmolarity. JNKK1 phosphorylates JNK and p38 but not the ERK MAP kinases. Active JNK and p38 phosphorylate multiple transcription factors that induce expression of genes, such as proinflammatory cytokines.

TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK/SAPK1
Applications Western Blot
Classification Monoclonal
Clone 37
Concentration 250μg/mL
Conjugate Unconjugated
Description p49 MAPK
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.