Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNK3/SAPK1b Rabbit anti-Human, Rat, Clone: C05T, MilliporeSigma™

Catalog No. 04893MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-893-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-893-MI Supplier MilliporeSigma Supplier No. 04893
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Detect JNK3/SAPK1b using this Anti-JNK3/SAPK1b Antibody, clone C05T validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK3/SAPK1b
Applications Western Blot
Classification Monoclonal
Clone C05T
Conjugate Unconjugated
Formulation Cultured supernantant in 0.05% sodium azide.
Gene Accession No. NM_002753.2
Host Species Rabbit
Immunogen GST fusion protein corresponding to full-length rat JNK3/ SAPK1b
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) EC 2.7.11.24, FLJ12099, FLJ33785, JNK3, JNK3A, MGC50974, OTTHUMP00000161180, OTTHUMP00000161182, OTTHUMP00000161183, PRKM10, p493F12, p54bSAPK
Target Species Human, Rat
Content And Storage −20°C for 12 months
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.