Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

jerky homolog (mouse), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016950 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-950 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-950 Supplier Abnova Corporation Supplier No. H00008629B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human JRK protein.

This gene is the human homolog of the mouse jerky gene. The encoded protein has similarity to several nuclear regulatory proteins, including centromere protein B, suggesting that it might function as a DNA-binding protein. Insertional inactivation of this gene in transgenic mice resulted in epileptic seizures. Childhood Absence Epilepsy (CAE) has been mapped to the same chromosomal location (8q24.3) as this gene, making this gene a strong candidate for CAE. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MASKPAAGKSRGEKRKRVVLTLKEKIDICTRLEKGESRKALMQEYNVGMSTLYDIRAHKAQLLRFFASSDSNKALEQRRTLHTPKLEHLDRVLYEWFLGKRSEGVPVSGPMLIEKAKDFYEQMQLTEPCVFSGGWLWRFKARHGIKKLDASSEKQSADHQAAEQFCAFFRSLAAEHGLSAEQVYNADETGLFWRCLPNPTPEGGAVPGPKQGKDRLTVLMCANATGSHRLKPLAIGKCSGPRAFKGIQHLPVAYKAQGNAWVDKEIFSDWFHHIFVPSVREHFRTIGLPEDSKAVLLLDSSRAHPQEAELVSSNVFTIFLPASVASLVQPMEQGIRRDFMRNFINPPVPLQGPHARYNMNDAIFSVACAWNAVPSHVFRRAWRKLWPSVAFAEGSSSEEELEAECFPVKPHNKSFAHILELVKEGSSCPGQLRQRQAASWGVAGREAEGGRPPAATSPAEVVWSSEKTPKADQDGRGDPGEGEEVAWEQAAVAFDAVLRFAERQPCFSAQEVGQLRALRAVFRSQQQVRRRRGALGAVVKVEALQEGPGGCGATAQSPLPCSSTAGDN

Specifications

Antigen jerky homolog (mouse)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human JRK protein.
Formulation PBS with no preservative; pH 7.4
Gene JRK
Gene Accession No. NM_003724.1
Gene Alias DKFZp686C24207/FLJ45729/JH8
Gene Symbols JRK
Host Species Mouse
Immunogen JRK (AAH43351.1, 1 a.a. ∼ 568 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8629
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.