Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
jumping translocation breakpoint (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human JTB protein.
Supplier: Abnova Corporation H00010899B02
Description
Sequence: MLAGAGRPGLPQGRHLCWLLCAFTLKLCQAEAPVQEEKLSASTSNLPCWLVEEFVVAEECSPCSNFRAKTTPECGPTGYVEKITCSSSKRNEFKSCRSALMEQRLFWKFEGAVVCVALIFACLVIIRQRQLDRKALEKVRKQIESISpecifications
jumping translocation breakpoint | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human JTB protein. | |
JTB | |
HJTB/HSPC222/PAR/hJT | |
Mouse | |
50 μL | |
Cell Adhesion | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_006694.2 | |
JTB | |
JTB (NP_006685.1, 1 a.a. ∼ 146 a.a) full-length human protein. | |
RUO | |
Yes | |
10899 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction