Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JTV1 gene, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016939 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-939 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-939 Supplier Abnova Corporation Supplier No. H00007965B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human JTV1 protein.

The JTV1 gene is located on chromosome 7p22 flanked by two genes, HRI and PMS2. JTV1 and HRI overlap slightly and are arranged in a tail-to-tail fashion. JTV1 and PMS2 are separated by approximately 200 base pairs and are arranged head-to-head. JTV1 is transcribed in the opposite direction compared to HRI and PMS2. The function of the JTV1 gene product is unknown. [provided by RefSeq

Sequence: MPMYQVKPYHGGGAPLRVELPTCMYRLPNVHGRSYGPAPGAGHVQEESNLSLQALESRQDDILKRLYELKAAVDGLSKMIQTPDADLDVTNIIQADEPTTLTTNALDLNSVLGKDYGALKDIVINANPASPPLSLLVLHRLLCEHFRVLSTVHTHSSVKSVPENLLKCFGEQNKKQPRQDYQLGFTLIWKNVPKTQMKFSIQTMCPIEGEGNIARFLFSLFGQKHNAVNATLIDSWVDIAIFQLKEGSSKEKAAVFRSMNSALGKSPWLAGNELTVADVVLWSVLQQIGGCSVTVPANVQRWMRSCENLAPFNTALKLLK

Specifications

Antigen JTV1 gene
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human JTV1 protein.
Formulation PBS with no preservative; pH 7.4
Gene JTV1
Gene Accession No. NM_006303.3
Gene Alias AIMP2/P38/PRO0992
Gene Symbols JTV1
Host Species Mouse
Immunogen JTV1 (NP_006294.2, 1 a.a. ∼ 320 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7965
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.