Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K+/Cl- Cotransporter (KCC2) Rabbit anti-Rat, Canine, Polyclonal, MilliporeSigma™

Catalog No. 07432MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-432-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-432-MI Supplier MilliporeSigma Supplier No. 07432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-K+/Cl-Cotransporter (KCC2) Antibody is validated for use in immunocytochemistry, western blotting for the detection of K+/Cl-Cotransporter (KCC2) with more than 15 product citations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen K+/Cl− Cotransporter (KCC2)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.014M Phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide and 30% glycerol. Liquid at −20°C.
Gene Symbols SLC12A5; KIAA1176; hKCC2; KCC2
Host Species Rabbit
Immunogen N-terminal His-tag fusion protein corresponding to residues 932-1043 of rat K+/Cl− Cotransporter (KCC2).
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_065759
Target Species Canine, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.