Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UFL1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526680
Description
This antibody is predicted to react with rat based on sequence homology.
Specifications
| UFL1 | |
| Polyclonal | |
| UFL1 | |
| KIAA0776, Maxer, NLBP, RCAD, RP3-393D12.1, E3 UFM1-protein ligase 1, LZAP-binding protein, Regulator of CDK5RAP3 and DDRGK1, novel LZAP-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23376 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| O94874 | |
| UFL1 | |
| KLH conjugated synthetic peptide between 426-455 amino acids from the central region of human K0776 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction