Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
K2P4.1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB983050UL
Description
Anti-K2P4.1 Antibody is an antibody against K2P4.1 for use in immunohistochemistry and western blotting.
Specifications
K2P4.1 | |
Polyclonal | |
Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.025% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
KCNK4; TRAAK; K2p4.1; TRAAK1 | |
Purified peptide from amino acids 343-359 of human K2P4.1 | |
50 μL | |
NM_033310.2 | |
Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction