Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

keratin 75, Mouse, Clone: 2E3, Abnova™

Catalog No. 89001505 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-505 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-505 Supplier Abnova Corporation Supplier No. H00009119M01A
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant K6HF.

This gene is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. This gene is expressed in the companion layer, upper germinative matrix region of the hair follicle, and medulla of the hair shaft. The encoded protein plays an essential role in hair and nail formation. Variations in this gene have been associated with the hair disorders pseudofolliculitis barbae (PFB) and loose anagen hair syndrome (LAHS). [provided by RefSeq

Sequence: TIQRVRAEEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWALLQEQGSRTVRQNLEPLFDSYTSELRRQLESITTERGRLEAELRNMQDVVEDFKVRYE

Specifications

Antigen keratin 75
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant K6HF.
Formulation ascites with no preservative
Gene KRT75
Gene Accession No. NM_004693
Gene Alias K6HF/KB18/PFB
Gene Symbols KRT75
Host Species Mouse
Immunogen K6HF (NP_004684, 141 a.a. ∼ 240 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9119
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.