Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kaiso Mouse anti-Canine, Chicken, Human, Hamster, Mouse, Monkey, Rat, Clone: 6F, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05659
Description
Detect Kaiso with Anti-Kaiso Antibody, clone 6F (Mouse Monoclonal Antibody), that has been shown to work in EMSA, immunoprecipitation, western blotting and immunohistochemistry.
Specifications
Kaiso | |
Monoclonal | |
Unconjugated | |
NM_006777 | |
Mouse | |
Protein G chromatography | |
Primary | |
−20°C for 24 months. | |
IgG1 |
Electromobility Shift Assay, Immunohistochemistry, Immunoprecipitation, Western Blot | |
6F | |
0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol. | |
ZBTB33, ZNF348, kaiso, ZNF-kaiso, WUGSC:H_DJ525N14.1, KAISO | |
Recombinant N-terminal GST and His-tagged fusion protein corresponding to residues 103-584 of murine Kaiso | |
200 μg | |
Canine, Chicken, Hamster, Human, Monkey, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction