Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kalirin Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07122MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-122-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-122-MI Supplier MilliporeSigma Supplier No. 07122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Kalirin Antibody is an antibody against Kalirin for use in immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kalirin
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide prior to the addition of glycerol to 30%
Gene Symbols KALRN; TRAD; Hs.8004; DUET; duo; Kalirin; FLJ16443; DUO; HAPIP
Host Species Rabbit
Immunogen GST fusion protein corresponding to residues 517-976 of rat Kalirin, cleaved with thrombin to remove the GST moiety; referred to as JH2582
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_032062
Target Species Mouse, Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.