Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein-3 (KLK3) (CT) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB10428MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10428MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10428MI Supplier MilliporeSigma Supplier No. AB10428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Kallikrein-3 (KLK3) (C-term) Antibody is validated for use in western blotting for the detection of Kallikrein-3 (KLK3) (C-term).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein-3 (KLK3) (CT)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in PBS with 0.09% NaN3.
Gene Symbols APS; EC 3.4.21.77; Gamma-seminoprotein ; KLK2A1; Kallikrein-3; PSA ; Semenogelase; Seminin; hK3
Host Species Rabbit
Immunogen This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human KLK3.
Purification Method Saturated Ammonium Sulfate (SAS)
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001639
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months, −20°C for long term. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.