Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-KAP-1/TRIM28, Polyclonal

Catalog No. ABE1859MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1859MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1859MI Supplier MilliporeSigma™ Supplier No. ABE1859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects KAP-1/TRIM28 Clone: in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting

Transcription intermediary factor 1-beta (UniProt Q13263; also known as E3 SUMO-protein ligase TRIM28, KAP-1, KRAB-associated protein 1, KRAB-interacting protein 1, KRIP-1, Nuclear corepressor KAP-1, Protein phosphatase 1 regulatory subunit 157, RING finger protein 96, TIF1-beta, Transcriptional intermediary factor 1-beta, Tripartite motif-containing 28, Tripartite motif-containing protein 28) is encoded by the TRIM28 (also known as KAP1, PPP1R157, RNF96, TF1B, TIF1B) gene (Gene ID 10155) in human. Kruppel-associated box-/KRAB-associated protein 1 (KAP-1) is a transcriptional co-repressor that selectively targets the KRAB domain of zinc finger protein-based transcription factors with its N-terminal tripartite RBCC (RING, b-box, coiled-coil) domain. KAP-1 uses its C-terminal PxVxL motif and the PHD-Bromo domain to recruit chromatin modifiers, such as heterochromatin protein 1 (HP-1), Histone-lysine N-methyltransferase SETDB1, nucleosome remodeling and deacetylation (NuRD) complex, and histone deacetylases (HDACs), for transcriptional repression. KAP-1 also plays an essential role in the DNA damage response (DDR). Upon DNA damage, KAP-1 is rapidly phosphorylated by Ataxia telangiectasia mutated (ATM) on Ser824, causing chromatin relaxation to facilitate DNA repair. In addition, SIRT1-mediated KAP-1 deacetylation is reported to stabilize KAP1 and 53BP1 interaction, resulting in an enhancement of NHEJ-mediated repair efficiency in a KAP-1 pSer824-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KAP-1/TRIM28
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody in serum with 0.05% sodium azide.
Gene Accession No. Q13263
Gene Symbols TRIM28, KAP1, RNF96, TIF1B
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the Bromo domain of human KAP-1/TRIM28.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005753
Test Specificity Expected to react with both spliced isoforms.
Target Species Human, Mouse
Content And Storage −20°C for one year
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.