Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KChIP2 potassium channel subunit Mouse anti-Mouse, Rat, Human, Clone: 60/73, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN371
Description
This Anti-KChIP2 potassium channel subunit Antibody, clone 60/73 is validated for use in western blotting and immunohistochemistry for the detection of KChIP2 potassium channel subunit.
Specifications
KChIP2 potassium channel subunit | |
Monoclonal | |
Unconjugated | |
NP_001029133 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
60/73 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Kcnip2, Kchip2 | |
GST-tagged recombinant protein corresponding to rat KChIP2 potassium channel subunit. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction