Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

potassium voltage-gated channel, Isk-related family, member 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017797 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-797 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-797 Supplier Abnova Corporation Supplier No. H00003753D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human KCNE1 protein.

The product of this gene belongs to the potassium channel KCNE family. Potassium ion channels are essential to many cellular functions and show a high degree of diversity, varying in their electrophysiologic and pharmacologic properties. This gene encodes a transmembrane protein known to associate with the product of the KVLQT1 gene to form the delayed rectifier potassium channel. Mutation in this gene are associated with both Jervell and Lange-Nielsen and Romano-Ward forms of long-QT syndrome. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq

Sequence: MILSNTTAVTPFLTKLWQETVQQGGNMSGLARRSPRSSDGKLEALYVLMVLGFFGFFTLGIMLSYIRSKKLEHSNDPFNVYIESDAWQEKDKAYVQARVLESYRSCYVVENHLAIEQPNTHLPETKPSP

Specifications

Antigen potassium voltage-gated channel, Isk-related family, member 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human KCNE1 protein.
Formulation PBS with no preservative; pH 7.4
Gene KCNE1
Gene Accession No. NM_000219.2
Gene Alias FLJ18426/FLJ38123/FLJ94103/ISK/JLNS/JLNS2/LQT2/5/LQT5/MGC33114/MinK
Gene Symbols KCNE1
Host Species Rabbit
Immunogen KCNE1 (NP_000210.2, 1 a.a. ∼ 129 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3753
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.