Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

potassium voltage-gated channel, subfamily G, member 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004811 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-811 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-811 Supplier Abnova Corporation Supplier No. H00170850A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant KCNG3.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member is a gamma subunit functioning as a modulatory molecule. Alternative splicing results in two transcript variants encoding distinct isoforms. [provided by RefSeq

Sequence: SRELLKDFPLRRVSRLHGCRSERDVLEVCDDYDRERNEYFFDRHSEAFGFILLYVRGHGKLRFAPRMCELSFYNEMIYWGLEGAHLEYCCQRRLDDRMS

Specifications

Antigen potassium voltage-gated channel, subfamily G, member 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant KCNG3.
Formulation 50% glycerol
Gene KCNG3
Gene Accession No. NM_133329
Gene Alias KV10.1/KV6.3
Gene Symbols KCNG3
Host Species Mouse
Immunogen KCNG3 (NP_579875, 23 a.a. ∼ 121 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 170850
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.