Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

potassium inwardly-rectifying channel, subfamily J, member 16, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004180 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-180 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-180 Supplier Abnova Corporation Supplier No. H00003773A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant KCNJ16.

Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, can form heterodimers with two other inward-rectifier type potassium channels. It may be involved in the regulation of fluid and pH balance. Three transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: KYYKVNCLQFEGSVEVYAPFCSAKQLDWKDQQLHIEKAPPVRESCTSDTKARRRSFSAVAIVSSCENPEETTTSATHEYRETPYQKALLTLNRISVES

Specifications

Antigen potassium inwardly-rectifying channel, subfamily J, member 16
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant KCNJ16.
Formulation 50% glycerol
Gene KCNJ16
Gene Accession No. NM_018658
Gene Alias BIR9/KIR5.1/MGC33717
Gene Symbols KCNJ16
Host Species Mouse
Immunogen KCNJ16 (NP_061128, 319 a.a. ∼ 416 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3773
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.