Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

potassium intermediate/small conductance calcium-activated channel, subfamily N, member 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-181 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-181 Supplier Abnova Corporation Supplier No. H00003782A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant KCNN3.

Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. The protein encoded by this gene is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits. This gene contains two CAG repeat regions in the coding sequence. It was thought that expansion of one or both of these repeats could lead to an increased susceptibility to schizophrenia or bipolar disorder, but studies indicate that this is probably not the case. This gene is a member of the KCNN family of potassium channel genes. Two transcript variants encoding two different isoforms have been found for this gene. One of the variants lacks the CAG repeat regions. [provided by RefSeq

Sequence: QAIHQLRSVKMEQRKLSDQANTLVDLSKMQNVMYDLITELNDRSEDLEKQIGSLESKLEHLTASFNSLPLLIADTLRQQQQQLLSAIIEARGVSVAVGTTHTPISDSPIG

Specifications

Antigen potassium intermediate/small conductance calcium-activated channel, subfamily N, member 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant KCNN3.
Formulation 50% glycerol
Gene KCNN3
Gene Accession No. NM_170782
Gene Alias KCa2.3/SK3/SKCA3/hSK3
Gene Symbols KCNN3
Host Species Mouse
Immunogen KCNN3 (NP_740752, 301 a.a. ∼ 410 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3782
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.