Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCTD15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520702
Description
A suggested positive control is Hela cell lysate. PA5-20702 can be used with blocking peptide PEP-0817.
Childhood and adult obesity in the United States and to a lesser extent the rest of the world has increased dramatically over the past decade. Both environmental and genetic factors are involved in the onset and progression of weight gain. Recently, the potassium channel KCTD15 was identified as a genetic loci associated with higher than normal body mass index (BMI) in humans along with genes such as GNPDA2, MTCH2, FTO, and TMEM18. Further studies on single nucleotide polymorphisms (SNPs) in non-diabetic and diabetic patients showed that FTO was most strongly associated with obesity while MTCH2 and GNPDA2 were still significantly associated with higher than normal BMI levels. At least two isoforms of KCTD15 are known to exist.
Specifications
KCTD15 | |
Polyclonal | |
Unconjugated | |
KCTD15 | |
BC031749; BTB/POZ domain-containing protein KCTD15; Kctd15; potassium channel tetramerisation domain containing 15; potassium channel tetramerization domain containing 15; potassium channel tetramerization domain-containing protein 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
233107, 499129, 79047 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8K0E1, Q96SI1 | |
KCTD15 | |
A 16 amino acid peptide near the carboxy terminus of human KCTD15. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction