Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-KDEL Clone: 10C3,

Catalog No. 42040050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
42-040-050UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-040-050UG Supplier MilliporeSigma™ Supplier No. 42040050UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects KDEL Clone: 10C3 in Canine, Drosophila, Guinea Pig, Hamster, Human, Monkey, Mouse, Porcine, Rabbit, Rat, Sheep, Yeast samples, and it is validated for Immunocytochemistry, Immunoprecipitation

Several resident proteins in the ER are involved in the proper assembly and folding of newly synthesized proteins. These include the glucose-regulated protein 94 (Grp94), BiP, which functions in protein oligomerization, and protein disulfide isomerase (PDI), which catalyzes the rearrangement of the disulfide bond. Grp94, BiP, and PDI share a carboxy-terminal sequence (Lys-Asp-Glu-Leu) with other resident ER proteins. The KDEL sequence is sufficient for the retention of a protein in the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KDEL
Applications Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 10C3
Conjugate Unconjugated
Formulation In PBS, 50% glycerol.
Host Species Mouse
Immunogen a synthetic peptide (SEKDEL) corresponding to amino acids 649-654 of rat Grp78, conjugated to KLH
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the Grp78 (BiP) and Grp94 proteins.
Target Species Canine, Drosophila, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Yeast
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.