Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

keratocan, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89018861 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-861 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-861 Supplier Abnova Corporation Supplier No. H00011081A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant KERA.

Keratan sulfate proteoglycans (KSPGs) are members of the small leucine-rich proteoglycan (SLRP) family. KSPGs, particularly keratocan, lumican (MIM 600616), and mimecan (MIM 602383), are important to the transparency of the cornea.[supplied by OMIM

Sequence: GLPSRGFDVSSILDLQLSHNQLTKVPRISAHLQHLHLDHNKIKSVNVSVICPSPSMLPAERDSFSYGPHLRYLRLDGNEIKPPIPMALMTCFRLLQAVI

Specifications

Antigen keratocan
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant KERA.
Formulation 50% glycerol
Gene KERA
Gene Accession No. NM_007035
Gene Alias CNA2/SLRR2B
Gene Symbols KERA
Host Species Mouse
Immunogen KERA (NP_008966, 253 a.a. ∼ 351 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11081
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.