Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratan Sulfate Mouse anti-Bovine, Canine, Human, Mouse, Pig, Sheep, Clone: EFG-11, MilliporeSigma™

Catalog No. MAB2022MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2022MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2022MI Supplier MilliporeSigma Supplier No. MAB2022
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Keratan Sulfate Antibody, clone EFG-11 is an antibody against Keratan Sulfate for use in ELISA, RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Keratan Sulfate
Applications ELISA, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone EFG-11
Conjugate Unconjugated
Formulation Ascites liquid, no preservative.
Gene Accession No. NM_002345.3
Gene Symbols LUM, SLRR2D, LDC, lumican
Host Species Mouse
Immunogen Human articular cartilage proteoglycan digested with chondroitinase ABC.
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Mouse, Pig, Sheep
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.